Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF1AX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_849416.5 | XP_854509.1 | 3 - 4 | 430 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |