Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022419386.1 | XP_022275094.1 | 1 - 2 | 50 | 110 | |
XM_848905.5 | XP_853998.2 | 1 - 2 | 87 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |