Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014109976.2 | XP_013965451.1 | 2 - 3 | 338 | 70 | |
XM_014109977.2 | XP_013965452.1 | 2 - 3 | 315 | 70 | ||
XM_014109978.2 | XP_013965453.1 | 2 - 3 | 339 | 70 | ||
XM_014109979.2 | XP_013965454.1 | 2 - 3 | 401 | 70 | ||
XM_014109980.2 | XP_013965455.1 | 2 - 3 | 414 | 70 | ||
XM_014109981.2 | XP_013965456.1 | 2 - 3 | 340 | 70 | ||
XM_848036.5 | XP_853129.1 | 2 - 3 | 268 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons |