Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145174.1 | NP_001138646.1 | 6 - 7 | 913 | 79 | |
XM_022424930.1 | XP_022280638.1 | 6 - 7 | 955 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |