Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145174.1 | NP_001138646.1 | 8 - 9 | 1213 | 54 | |
XM_022424930.1 | XP_022280638.1 | 8 - 9 | 1255 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |