Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLYRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_849945.4 | XP_855038.1 | 1 - 2 | 320 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons |