Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_848549.5 | XP_853642.1 | 2 - 3 | 450 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |