Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005635632.2 | XP_005635689.1 | 1004 | 148 | ||
XM_014107596.2 | XP_013963071.1 | 1067 | 148 | |||
XM_014107597.2 | XP_013963072.1 | 1055 | 148 | |||
XM_022409784.1 | XP_022265492.1 | 1023 | 148 | |||
XM_534550.6 | XP_534550.2 | 991 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris |