Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142670.1 | NP_001136142.1 | 1 - 2 | 230 | 86 | |
GenBank mRNA | AB467286.1 | - | 1 - 2 | 230 | 86 | |
AB467287.1 | - | 1 - 2 | 160 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |