Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005625524.3 | XP_005625581.1 | 734 | 62 | ||
XM_022424071.1 | XP_022279779.1 | 686 | 62 | |||
XM_022424072.1 | XP_022279780.1 | 729 | 62 | |||
XM_022424074.1 | XP_022279782.1 | 694 | 62 | |||
XM_022424075.1 | XP_022279783.1 | 698 | 62 | |||
XM_022424076.1 | XP_022279784.1 | 779 | 62 | |||
XM_022424077.1 | XP_022279785.1 | 722 | 62 | |||
XM_022424078.1 | XP_022279786.1 | 654 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris |