Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5PO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638881.3 | XP_005638938.1 | 5 - 6 | 540 | 142 |
Gene Symbol: ATP5O | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638862.3 | XP_005638919.1 | 5 - 6 | 540 | 142 | |
GenBank mRNA | CV999268.1 | - | 5 - 6 | 462 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Probe spans exons |