Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COASY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624450.3 | XP_005624507.1 | 1644 | 71 | ||
XM_005624451.2 | XP_005624508.1 | 1306 | 71 | |||
XM_537637.6 | XP_537637.2 | 1372 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris |