Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014110994.2 | XP_013966469.1 | 1 - 2 | 295 | 107 | |
XM_022415180.1 | XP_022270888.1 | 1 - 2 | 292 | 107 | ||
XM_022415181.1 | XP_022270889.1 | 1 - 2 | 291 | 107 | ||
XM_022415182.1 | XP_022270890.1 | 1 - 2 | 290 | 107 | ||
XM_022415183.1 | XP_022270891.1 | 1 - 2 | 291 | 107 | ||
XM_845032.4 | XP_850125.1 | 1 - 2 | 285 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons |