Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014119564.2 | XP_013975039.1 | 7 - 8 | 2392 | 96 | |
XM_014119565.2 | XP_013975040.1 | 7 - 8 | 1927 | 96 | ||
XM_022427982.1 | XP_022283690.1 | 7 - 8 | 2303 | 96 | ||
XM_845204.5 | XP_850297.1 | 7 - 8 | 2302 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Probe spans exons |