Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252337.1 | NP_001239266.1 | 323 | 116 | ||
XM_005619533.3 | XP_005619590.1 | 868 | 116 | |||
XM_005619534.3 | XP_005619591.2 | 391 | 116 | |||
XM_005619537.3 | XP_005619594.2 | 297 | 116 | |||
XM_014113205.2 | XP_013968680.1 | 461 | 116 | |||
GenBank mRNA | DN330290.1 | - | 323 | 116 | ||
DN348014.1 | - | 222 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris |