Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAZAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005636814.2 | XP_005636871.1 | 3 - 4 | 509 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |