Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005617615.2 | XP_005617672.1 | 3 - 4 | 740 | 152 | |
XM_022411818.1 | XP_022267526.1 | 3 - 4 | 626 | 152 | ||
XM_849499.5 | XP_854592.3 | 3 - 4 | 611 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |