Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_846608.5 | XP_851701.1 | 2 - 3 | 632 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |