Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002968.1 | NP_001002968.1 | 4 - 5 | 762 | 90 | |
GenBank mRNA | AJ581082.1 | - | 4 - 5 | 762 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |