Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005623314.3 | XP_005623371.1 | 4 - 5 | 4336 | 190 | |
XM_845275.5 | XP_850368.1 | 4 - 5 | 4339 | 190 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |