Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_534901.6 | XP_534901.2 | 4 - 5 | 745 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |