Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005631904.3 | XP_005631961.1 | 4 - 5 | 583 | 180 | |
XM_541001.5 | XP_541001.2 | 4 - 5 | 704 | 180 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |