Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252264.1 | NP_001239193.1 | 1 | 399 | 93 | |
NM_001252265.1 | NP_001239194.1 | 2 | 378 | 93 | ||
GenBank mRNA | DN370387.1 | - | 2 | 378 | 93 | |
DN371621.1 | - | 354 | 93 | |||
DN865199.1 | - | 1 | 113 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |