Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022423258.1 | XP_022278966.1 | 4 - 5 | 1230 | 71 | |
XM_847666.5 | XP_852759.2 | 4 - 5 | 1231 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons |