Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003640046.4 | XP_003640094.1 | 3 | 1653 | 94 | |
XM_005638333.3 | XP_005638390.1 | 4 | 1129 | 94 | ||
XM_005638334.3 | XP_005638391.1 | 4 | 1157 | 94 | ||
XM_005638335.3 | XP_005638392.1 | 4 | 779 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Amplicon spans exons and probe does not span exons |