Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPF38B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022420382.1 | XP_022276090.1 | 6 - 7 | 843 | 150 | |
XM_849600.5 | XP_854693.1 | 4 - 5 | 639 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |