Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK1IP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005623420.3 | XP_005623477.1 | 3 | 198 | 122 | |
XM_005623421.2 | XP_005623478.1 | 3 | 199 | 122 | ||
XM_022422023.1 | XP_022277731.1 | 3 | 197 | 122 | ||
XM_022422024.1 | XP_022277732.1 | 3 | 399 | 122 | ||
XM_022422025.1 | XP_022277733.1 | 2 | 295 | 122 | ||
XM_537452.5 | XP_537452.2 | 3 | 317 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |