Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005635672.3 | XP_005635729.1 | 3 | 587 | 128 | |
XM_543223.6 | XP_543223.2 | 3 | 586 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Amplicon spans exons and probe does not span exons |