Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADRA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195843.1 | NP_001182772.1 | 1 | 845 | 59 | |
XM_005635607.3 | XP_005635664.1 | 3 | 1902 | 59 | ||
XM_022409542.1 | XP_022265250.1 | 2 | 2152 | 59 | ||
XM_022409544.1 | XP_022265252.1 | 2 | 2156 | 59 | ||
XM_022409545.1 | XP_022265253.1 | 2 | 2154 | 59 | ||
XM_022409546.1 | XP_022265254.1 | 2 | 2163 | 59 | ||
GenBank mRNA | AF068283.1 | - | 1 | 846 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |