Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048125.1 | NP_001041590.1 | 7 | 885 | 190 | |
XM_005641023.3 | XP_005641080.1 | 7 | 1032 | 190 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Amplicon spans exons and probe does not span exons |