Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005632592.3 | XP_005632649.1 | 3 | 375 | 122 | |
XM_014122342.2 | XP_013977817.1 | 5 | 2434 | 122 | ||
XM_846287.5 | XP_851380.1 | 4 | 693 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |