Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S1PR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314114.1 | NP_001301043.1 | 1 | 630 | 125 | |
XM_005621856.3 | XP_005621913.1 | 2 | 877 | 125 | ||
XM_005621857.3 | XP_005621914.1 | 2 | 1024 | 125 | ||
XM_005621858.3 | XP_005621915.1 | 2 | 793 | 125 | ||
XM_547259.6 | XP_547259.2 | 2 | 1023 | 125 | ||
GenBank mRNA | KF841603.1 | - | 1 | 630 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |