Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022416916.1 | XP_022272624.1 | 2 - 3 | 882 | 69 | |
XM_022416917.1 | XP_022272625.1 | 4 - 5 | 897 | 69 | ||
XM_022416918.1 | XP_022272626.1 | 5 - 6 | 1091 | 69 | ||
XM_022416920.1 | XP_022272628.1 | 2 - 3 | 688 | 69 | ||
XM_545846.6 | XP_545846.2 | 2 - 3 | 688 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons |