Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNIPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_540308.6 | XP_540308.3 | 4 | 805 | 175 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Amplicon spans exons and probe does not span exons |