Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003431935.4 | XP_003431983.1 | 3 | 577 | 102 | |
XM_005628728.3 | XP_005628785.1 | 3 | 622 | 102 | ||
XM_532518.6 | XP_532518.2 | 3 | 623 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |