Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRELP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005640760.3 | XP_005640817.1 | 2 | 893 | 94 | |
XM_545678.6 | XP_545678.2 | 2 | 902 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |