Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022410513.1 | XP_022266221.1 | 3 | 1082 | 141 | |
XM_022410514.1 | XP_022266222.1 | 3 | 1564 | 141 | ||
XM_022410515.1 | XP_022266223.1 | 2 | 1018 | 141 | ||
XM_849500.5 | XP_854593.2 | 2 | 1528 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |