Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005641408.3 | XP_005641465.1 | 1 | 867 | 162 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |