Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_846508.4 | XP_851601.2 | 706 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris |