Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PA2G4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_531629.5 | XP_531629.2 | 1207 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |