Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287118.1 | NP_001274047.1 | 3 | 351 | 181 | |
XM_022418006.1 | XP_022273714.1 | 3 | 378 | 181 | ||
GenBank mRNA | CV998536.1 | - | 3 | 342 | 181 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |