Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFE2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624613.3 | XP_005624670.1 | 1365 | 170 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris |