Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003357.2 | NP_001003357.2 | 8 - 9 | 756 | 84 | |
NM_001313818.1 | NP_001300747.1 | 7 - 8 | 649 | 84 | ||
GenBank mRNA | AY283372.1 | - | 8 - 9 | 607 | 84 | |
DN394227.1 | - | 8 - 9 | 766 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Endogenous Controls Detect Genomic DNA Recommended Assay Canis familiaris Inventoried Probe spans exons |