Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADRA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195843.1 | NP_001182772.1 | 2 | 1276 | 159 | |
XM_005635607.3 | XP_005635664.1 | 4 | 2333 | 159 | ||
XM_022409542.1 | XP_022265250.1 | 3 | 2583 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |