Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005640842.2 | XP_005640899.1 | 2 | 384 | 119 | |
XM_545788.6 | XP_545788.2 | 1 | 608 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |