Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003215.2 | NP_001003215.2 | 26 - 27 | 3644 | 100 | |
XM_014118873.2 | XP_013974348.1 | 27 - 28 | 3817 | 100 | ||
XM_014118874.2 | XP_013974349.1 | 27 - 28 | 3747 | 100 | ||
GenBank mRNA | AF045016.1 | - | 27 - 28 | 3710 | 100 | |
AF536758.1 | - | 26 - 27 | 3660 | 100 | ||
AJ419568.1 | - | 26 - 27 | 3646 | 100 | ||
AY582533.1 | - | 26 - 27 | 3644 | 100 | ||
DN877923.1 | - | 1 - 2 | 636 | 100 | ||
DQ068953.1 | - | 26 - 27 | 3644 | 100 | ||
FJ617477.1 | - | 26 - 27 | 3644 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Inventoried Probe spans exons |