Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005641577.3 | XP_005641634.1 | 3 - 4 | 520 | 151 | |
XM_005641578.3 | XP_005641635.1 | 3 - 4 | 520 | 151 | ||
XM_005641580.3 | XP_005641637.1 | 3 - 4 | 343 | 151 | ||
XM_014111833.2 | XP_013967308.1 | 3 - 4 | 520 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons |