Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165878.1 | NP_001159350.1 | 3 | 295 | 129 | |
GenBank mRNA | AB514445.1 | - | 3 | 295 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Inventoried Both primers and probe map within a single exon |