Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639880.4 | XP_003639928.1 | 2 - 3 | 1306 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |