Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620395.3 | XP_005620452.1 | 1508 | 63 | ||
XM_843797.5 | XP_848890.1 | 1494 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris |